LIF112851544 Life Chemicals Inc.

3-(5-Chloro-1H-benzoimidazol-2-yl)-propionic acid

  • MDL number MFCD06753442
  • Cas Number : 82138-56-5
  • Molecular Weight : 224.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(5-chloro-1H-benzimidazol-2-yl)propanoic acid
Inchi Code 1S/C10H9ClN2O2/c11-6-1-2-7-8(5-6)13-9(12-7)3-4-10(14)15/h1-2,5H,3-4H2,(H,12,13)(H,14,15)
Inchi Key LPODEKMPICKOID-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart