LIF117311854 Life Chemicals Inc.

7-Mercapto-3-methyl-[1,3,4]thiadiazolo[2,3-c][1,2,4]triazin-4-one pyridinium salt

  • MDL number MFCD09235450
  • Cas Number : 94350-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart