LIF117312529 Life Chemicals Inc.

6-Oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00139896
  • Cas Number : 338783-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart