LIF117312726 Life Chemicals Inc.

6-Methyl-3-thioxo-3,4-dihydro-2H-[1,2,4]triazin-5-one

  • MDL number MFCD00006458
  • Cas Number : 615-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart