LIF117643550 Life Chemicals Inc.

2-Methyl-5-(2-thienylcarbonyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09743080
  • Cas Number : 941868-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart