LIF117643621 Life Chemicals Inc.

3-(1H-indol-3-yl)-1H-pyrazol-5-amine

  • MDL number MFCD09743154
  • Cas Number : 1040724-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart