LIF117643717 Life Chemicals Inc.

5-(Aminomethyl)-3-(4-chlorophenyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD09743251
  • Cas Number : 1177315-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart