LIF117643721 Life Chemicals Inc.

2,3-Dihydro-1H-indole-2-carboxylic acid amide

  • MDL number MFCD09743255
  • Cas Number : 108906-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart