LIF117643785 Life Chemicals Inc.

Indol-1-yl-acetic acid

  • MDL number MFCD00047262
  • Cas Number : 24297-59-4
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart