LIF117643801 Life Chemicals Inc.

(1{H}-indol-3-ylthio)acetic acid

  • MDL number MFCD00457137
  • Cas Number : 54466-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart