LIF117643814 Life Chemicals Inc.

7-Bromo-1,4-dihydro-pyrido[2,3-b]pyrazine-2,3-dione

  • MDL number MFCD00460131
  • Cas Number : 168123-82-8
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart