LIF117643918 Life Chemicals Inc.

[4-(2-Piperidin-1-ylethoxy)phenyl]boronic acid

  • MDL number MFCD09743123
  • Cas Number : 710348-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart