LIF127982978 Life Chemicals Inc.

3-Allyl-4-hydroxy-5-methoxy-benzaldehyde

  • MDL number MFCD01590282
  • Cas Number : 20240-58-8
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart