LIF127983070 Life Chemicals Inc.

[1,8]Naphthyridine-4-carboxylic acid

  • MDL number MFCD05664461
  • Cas Number : 99066-71-4
  • Molecular Weight : 174.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart