LIF127983071 Life Chemicals Inc.

7-Chloro-2,4-dimethyl-[1,8]naphthyridine

  • MDL number MFCD00206716
  • Cas Number : 77223-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart