LIF127983076 Life Chemicals Inc.

7-Methyl-2,3,4,5-tetrahydro-1-benzothiepin-5-ol 1,1-dioxide

  • MDL number MFCD09283623
  • Cas Number : 924859-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart