LIF127983758 Life Chemicals Inc.

2-(ethylamino)benzoic acid

  • MDL number MFCD00020248
  • Cas Number : 89-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart