LIF127987831 Life Chemicals Inc.

7-Chloro-1-methyl-1H-indole-2,3-dione

  • MDL number MFCD08755446
  • Cas Number : 63220-48-4
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart