LIF127987838 Life Chemicals Inc.

1-Methyloctahydroisoquinolin-4a(2H)-ol

  • MDL number MFCD05856377
  • Cas Number : 81562-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart