LIF127996114 Life Chemicals Inc.

5-(Chloromethyl)-1-(4-fluorophenyl)-1H-tetrazole

  • MDL number MFCD08548459
  • Cas Number : 912906-94-6
  • Molecular Weight : 212.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(chloromethyl)-1-(4-fluorophenyl)-1H-tetraazole
Inchi Code 1S/C8H6ClFN4/c9-5-8-11-12-13-14(8)7-3-1-6(10)2-4-7/h1-4H,5H2
Inchi Key FIGNQIBUPZMKSM-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart