LIF156487064 Life Chemicals Inc.

8-Bromo-[1,6]naphthyridine

  • MDL number MFCD00234384
  • Cas Number : 17965-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart