LIF156487065 Life Chemicals Inc.

1,6-Naphthyridine-8-carboxylic acid

  • MDL number MFCD09863867
  • Cas Number : 362606-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart