LIF156487150 Life Chemicals Inc.

2,3-Dimethyl-1H-indole-7-carboxylic acid

  • MDL number MFCD00090259
  • Cas Number : 103986-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart