LIF156505090 Life Chemicals Inc.

3-Bromo-2,4-dimethyl-1,8-naphthyridine

  • MDL number MFCD09863865
  • Cas Number : 1019108-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart