LIF156505257 Life Chemicals Inc.

2-(2-Amino-1,3-thiazol-4-yl)-N-methylacetamide

  • MDL number MFCD09864126
  • Cas Number : 640768-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart