LIF156505264 Life Chemicals Inc.

2-amino-N-methyl-1,3-thiazole-4-carboxamide

  • MDL number MFCD09864133
  • Cas Number : 827588-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart