LIF156505590 Life Chemicals Inc.

2-(5-fluoro-2-methyl-1H-indol-3-yl)acetic acid

  • MDL number MFCD02664389
  • Cas Number : 71987-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart