LIF156505630 Life Chemicals Inc.

2-Amino-4-phenyl-thiophene-3-carboxylic acid methyl ester

  • MDL number MFCD01050444
  • Cas Number : 67171-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart