LIF156505712 Life Chemicals Inc.

(4-Fluoro-benzenesulfonylamino)-acetic acid

  • MDL number MFCD02091617
  • Cas Number : 13029-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart