LIF156505765 Life Chemicals Inc.

4-Tetrazol-1-yl-phenol

  • MDL number MFCD03490124
  • Cas Number : 64001-11-2
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart