LIF156505791 Life Chemicals Inc.

2-(2-Amino-1,3-thiazol-4-yl)acetamide

  • MDL number MFCD07689461
  • Cas Number : 220041-33-8
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart