LIF156506068 Life Chemicals Inc.

(1,3-dimethyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD02055709
  • Cas Number : 400756-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart