LIF156506330 Life Chemicals Inc.

5,7-difluoro-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD09965712
  • Cas Number : 1019111-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart