LIF156506508 Life Chemicals Inc.

1-(3-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD06001923
  • Cas Number : 714207-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart