LIF156506517 Life Chemicals Inc.

1,2,5-thiadiazole-3-carboxylic acid

  • MDL number MFCD01334979
  • Cas Number : 13368-86-0
  • Molecular Weight : 130.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart