LIF156506549 Life Chemicals Inc.

1-methyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD04968667
  • Cas Number : 288148-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart