LIF156506731 Life Chemicals Inc.

3-(1-methyl-1H-pyrazol-4-yl)propanoic acid

  • MDL number MFCD04969921
  • Cas Number : 796845-56-2
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart