LIF156506934 Life Chemicals Inc.

5,6,7,8-Tetrahydro-naphthalene-2-sulfonyl chloride

  • MDL number MFCD06655218
  • Cas Number : 61551-49-3
  • Molecular Weight : 230.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart