LIF159318528 Life Chemicals Inc.

1-(2-Chlorobenzyl)-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD01935944
  • Cas Number : 4399-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart