LIF159321312 Life Chemicals Inc.

Potassium 5-pyridin-4-yl-1,3,4-oxadiazole-2-thiolate

  • MDL number MFCD04442758
  • Cas Number : 1024183-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart