LIF159730834 Life Chemicals Inc.

(1E)-N'-Hydroxy-3-(2-methyl-1H-benzimidazol-1-yl)propanimidamide

  • MDL number MFCD08436506
  • Cas Number : 537696-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart