LIF159730838 Life Chemicals Inc.

7-Hydroxy-5-methoxy-2-phenyl-4H-chromen-4-one

  • MDL number MFCD09029062
  • Cas Number : 33554-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart