LIF159730839 Life Chemicals Inc.

4-Hydroxy-6-methyl-1-benzofuran-3(2H)-one

  • MDL number MFCD09259072
  • Cas Number : 929339-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart