LIF159730840 Life Chemicals Inc.

4,7-Dihydroxy-3-phenyl-2H-chromen-2-one

  • MDL number MFCD01731806
  • Cas Number : 19225-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart