LIF159730841 Life Chemicals Inc.

6-Hydroxy-7-methyl-1-benzofuran-3(2H)-one

  • MDL number MFCD06496433
  • Cas Number : 21861-22-3
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart