LIF159730942 Life Chemicals Inc.

4-(2-Amino-1,3-thiazol-4-yl)-2-methylbenzene-1,3-diol

  • MDL number MFCD09259106
  • Cas Number : 929339-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart