LIF159730943 Life Chemicals Inc.

(4-Aminophenyl)(1-benzofuran-2-yl)methanone

  • MDL number MFCD09259109
  • Cas Number : 92789-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart