LIF159730982 Life Chemicals Inc.

4-Amino-8-chloroquinoline-3-carboxylic acid

  • MDL number MFCD10037652
  • Cas Number : 955328-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart