LIF159730989 Life Chemicals Inc.

6-Chloroquinolin-4-amine

  • MDL number MFCD06658304
  • Cas Number : 20028-60-8
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart