LIF161662813 Life Chemicals Inc.

6-Hydroxy-2-(1-methylethylidene)-1-benzofuran-3(2H)-one

  • MDL number MFCD06494097
  • Cas Number : 88281-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart